Anti-APP Mouse Monoclonal Antibody [clone: 2B10]

Supplier: OriGene
ORIGCF500973EA 749 EUR
ORIGCF500973
Anti-APP Mouse Monoclonal Antibody [clone: 2B10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:50

Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: 2B10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR