Anti-MCL1 Mouse Monoclonal Antibody [clone: 3B4]

Supplier: OriGene
ORIGCF500988EA 749 EUR
ORIGCF500988
Anti-MCL1 Mouse Monoclonal Antibody [clone: 3B4]
Antibodies
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: MCL1
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR