Anti-KLHL2 Mouse Monoclonal Antibody [clone: 1G7]

Supplier: OriGene
ORIGCF501617EA 749 EUR
ORIGCF501617
Anti-KLHL2 Mouse Monoclonal Antibody [clone: 1G7]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: KLHL2
Clonality: Monoclonal
Clone: 1G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR