Anti-GNB2L1 Mouse Monoclonal Antibody [clone: 2D8]

Supplier: OriGene
ORIGCF502717EA 749 EUR
ORIGCF502717
Anti-GNB2L1 Mouse Monoclonal Antibody [clone: 2D8]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: GNB2L1
Clonality: Monoclonal
Clone: 2D8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR