Anti-AK5 Mouse Monoclonal Antibody [clone: 2D3]

Supplier: OriGene
ORIGCF501649EA 770 EUR
ORIGCF501649
Anti-AK5 Mouse Monoclonal Antibody [clone: 2D3]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: AK5
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR