Anti-RABL2A Mouse Monoclonal Antibody [clone: 2G10]

Supplier: OriGene
ORIGCF502022EA 749 EUR
ORIGCF502022
Anti-RABL2A Mouse Monoclonal Antibody [clone: 2G10]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: RABL2A
Clonality: Monoclonal
Clone: 2G10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR