Anti-HDAC6 Mouse Monoclonal Antibody [clone: 4C5]

Supplier: OriGene
ORIGCF502030EA 749 EUR
ORIGCF502030
Anti-HDAC6 Mouse Monoclonal Antibody [clone: 4C5]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunochemistry: 1:150

Type: Primary
Antigen: HDAC6
Clonality: Monoclonal
Clone: 4C5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR