Anti-IVD Mouse Monoclonal Antibody [clone: 1F1]

Supplier: OriGene
ORIGCF501670EA 770 EUR
ORIGCF501670
Anti-IVD Mouse Monoclonal Antibody [clone: 1F1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: IVD
Clonality: Monoclonal
Clone: 1F1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR