Anti-GUK1 Mouse Monoclonal Antibody [clone: 1D7]

Supplier: OriGene
ORIGCF502659EA 770 EUR
ORIGCF502659
Anti-GUK1 Mouse Monoclonal Antibody [clone: 1D7]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: GUK1
Clonality: Monoclonal
Clone: 1D7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR