Imprimer

Anti-SULT1C2 Mouse Monoclonal Antibody [clone: 5A4]

Fournisseur: OriGene
ORIGCF502665EA 749 EUR
ORIGCF502665
Anti-SULT1C2 Mouse Monoclonal Antibody [clone: 5A4]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: SULT1C2
Clonality: Monoclonal
Clone: 5A4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR