Anti-NPR3 Mouse Monoclonal Antibody [clone: 11H6]

Supplier: OriGene
ORIGCF501080EA 749 EUR
ORIGCF501080
Anti-NPR3 Mouse Monoclonal Antibody [clone: 11H6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: NPR3
Clonality: Monoclonal
Clone: 11H6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR