Anti-ARHGAP25 Mouse Monoclonal Antibody [clone: 12G4]

Supplier: OriGene
ORIGCF501745EA 749 EUR
ORIGCF501745
Anti-ARHGAP25 Mouse Monoclonal Antibody [clone: 12G4]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ARHGAP25
Clonality: Monoclonal
Clone: 12G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR