Anti-PYCRL Mouse Monoclonal Antibody [clone: 5F7]

Supplier: OriGene
ORIGCF502121EA 749 EUR
ORIGCF502121
Anti-PYCRL Mouse Monoclonal Antibody [clone: 5F7]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PYCRL
Clonality: Monoclonal
Clone: 5F7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR