Anti-STAT5A Mouse Monoclonal Antibody [clone: 9F7]

Supplier: OriGene
ORIGCF502736EA 749 EUR
ORIGCF502736
Anti-STAT5A Mouse Monoclonal Antibody [clone: 9F7]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: STAT5A
Clonality: Monoclonal
Clone: 9F7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR