Anti-PRKAR1B Mouse Monoclonal Antibody [clone: 2A3]

Supplier: OriGene
ORIGCF502750EA 770 EUR
ORIGCF502750
Anti-PRKAR1B Mouse Monoclonal Antibody [clone: 2A3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PRKAR1B
Clonality: Monoclonal
Clone: 2A3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR