Anti-DNTTIP1 Mouse Monoclonal Antibody [clone: 4D1]

Supplier: OriGene
ORIGCF501131EA 749 EUR
ORIGCF501131
Anti-DNTTIP1 Mouse Monoclonal Antibody [clone: 4D1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: DNTTIP1
Clonality: Monoclonal
Clone: 4D1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR