Anti-ALDH3A1 Mouse Monoclonal Antibody [clone: 1C6]

Supplier: OriGene
ORIGCF501139EA 749 EUR
ORIGCF501139
Anti-ALDH3A1 Mouse Monoclonal Antibody [clone: 1C6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: ALDH3A1
Clonality: Monoclonal
Clone: 1C6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR