Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: 5D4]

Supplier: OriGene
ORIGCF501753EA 770 EUR
ORIGCF501753
Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: 5D4]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: PIK3AP1
Clonality: Monoclonal
Clone: 5D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR