Anti-AKT3 Mouse Monoclonal Antibody [clone: 9B2]

Supplier: OriGene
ORIGCF500001EA 749 EUR
ORIGCF500001
Anti-AKT3 Mouse Monoclonal Antibody [clone: 9B2]
Antibodies
Recommended Dilutions:Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100; FC: 1:100

Type: Primary
Antigen: AKT3
Clonality: Monoclonal
Clone: 9B2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR