Anti-MYC Mouse Monoclonal Antibody [clone: 1A6]

Supplier: OriGene
ORIGCF500002EA 749 EUR
ORIGCF500002
Anti-MYC Mouse Monoclonal Antibody [clone: 1A6]
Antibodies
Recommended Dilutions:Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; FC: 1:100

Type: Primary
Antigen: MYC
Clonality: Monoclonal
Clone: 1A6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR