Anti-HHex Mouse Monoclonal Antibody [clone: 2H7]

Supplier: OriGene
ORIGCF500027EA 749 EUR
ORIGCF500027
Anti-HHex Mouse Monoclonal Antibody [clone: 2H7]
Antibodies
Recommended Dilutions: Western Blot: 1:50-1:100; Immunofluorescence: 1:100

Type: Primary
Antigen: HHex
Clonality: Monoclonal
Clone: 2H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR