Anti-PDX1 Mouse Monoclonal Antibody [clone: 2A12]

Supplier: OriGene
ORIGCF500038EA 749 EUR
ORIGCF500038
Anti-PDX1 Mouse Monoclonal Antibody [clone: 2A12]
Antibodies
Recommended Dilutions: Western Blot: 1:20000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100

Type: Primary
Antigen: PDX1
Clonality: Monoclonal
Clone: 2A12
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR