Anti-SHH Mouse Monoclonal Antibody [clone: 3A2]

Supplier: OriGene
ORIGCF500040EA 749 EUR
ORIGCF500040
Anti-SHH Mouse Monoclonal Antibody [clone: 3A2]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: SHH
Clonality: Monoclonal
Clone: 3A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR