Anti-AFP Mouse Monoclonal Antibody [clone: 1B3]

Supplier: OriGene
ORIGCF502170EA 749 EUR
ORIGCF502170
Anti-AFP Mouse Monoclonal Antibody [clone: 1B3]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: 1B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR