Anti-LDHA Mouse Monoclonal Antibody [clone: 5G10]
Supplier: OriGene
ORIGCF500558EA 749 EUR
ORIGCF500558
Anti-LDHA Mouse Monoclonal Antibody [clone: 5G10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2?g/500?l
Type: Primary
Antigen: LDHA
Clonality: Monoclonal
Clone: 5G10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human