Anti-TUBB3 Mouse Monoclonal Antibody [clone: 5H2]

Supplier: OriGene
ORIGCF500047EA 749 EUR
ORIGCF500047
Anti-TUBB3 Mouse Monoclonal Antibody [clone: 5H2]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000

Type: Primary
Antigen: TUBB3
Clonality: Monoclonal
Clone: 5H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR