Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 1F3]

Supplier: OriGene
ORIGCF500048EA 749 EUR
ORIGCF500048
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 1F3]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50

Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone: 1F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR