Anti-EPCAM Mouse Monoclonal Antibody [clone: 2G1]

Supplier: OriGene
ORIGCF500054EA 749 EUR
ORIGCF500054
Anti-EPCAM Mouse Monoclonal Antibody [clone: 2G1]
Antibodies
Recommended Dilutions: Western Blot: 1:200-1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: 2G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR