Anti-PACSIN3 Mouse Monoclonal Antibody [clone: 4A7]

Supplier: OriGene
ORIGCF502215EA 770 EUR
ORIGCF502215
Anti-PACSIN3 Mouse Monoclonal Antibody [clone: 4A7]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PACSIN3
Clonality: Monoclonal
Clone: 4A7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR