Anti-KDM4C Mouse Monoclonal Antibody [clone: 4B1]

Supplier: OriGene
ORIGCF500587EA 749 EUR
ORIGCF500587
Anti-KDM4C Mouse Monoclonal Antibody [clone: 4B1]
Antibodies
Recommended Dilutions: Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: KDM4C
Clonality: Monoclonal
Clone: 4B1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR