Anti-IGF2BP2 Mouse Monoclonal Antibody [clone: 5G3]

Supplier: OriGene
ORIGCF501268EA 749 EUR
ORIGCF501268
Anti-IGF2BP2 Mouse Monoclonal Antibody [clone: 5G3]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: IGF2BP2
Clonality: Monoclonal
Clone: 5G3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR