Anti-NXNL2 Mouse Monoclonal Antibody [clone: 2H8]

Supplier: OriGene
ORIGCF501278EA 749 EUR
ORIGCF501278
Anti-NXNL2 Mouse Monoclonal Antibody [clone: 2H8]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NXNL2
Clonality: Monoclonal
Clone: 2H8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR