Anti-KHK Mouse Monoclonal Antibody [clone: 4E9]

Supplier: OriGene
ORIGCF501284EA 749 EUR
ORIGCF501284
Anti-KHK Mouse Monoclonal Antibody [clone: 4E9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: KHK
Clonality: Monoclonal
Clone: 4E9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR