Anti-NTF3 Mouse Monoclonal Antibody [clone: 5C7]

Supplier: OriGene
ORIGCF500115EA 749 EUR
ORIGCF500115
Anti-NTF3 Mouse Monoclonal Antibody [clone: 5C7]
Antibodies
Recommended Dilutions: Western Blot: 1:5000; Immunochemistry: 1:50

Type: Primary
Antigen: NTF3
Clonality: Monoclonal
Clone: 5C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR