Anti-Nog Mouse Monoclonal Antibody [clone: 1C1]

Supplier: OriGene
ORIGCF500116EA 749 EUR
ORIGCF500116
Anti-Nog Mouse Monoclonal Antibody [clone: 1C1]
Antibodies
Recommended Dilutions: Western Blot: 1:40000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: Nog
Clonality: Monoclonal
Clone: 1C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR