Anti-ID2 Mouse Monoclonal Antibody [clone: 9A8]

Supplier: OriGene
ORIGCF500199EA 749 EUR
ORIGCF500199
Anti-ID2 Mouse Monoclonal Antibody [clone: 9A8]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: ID2
Clonality: Monoclonal
Clone: 9A8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR