Anti-MSI1 Mouse Monoclonal Antibody [clone: 2E9]

Supplier: OriGene
ORIGCF502254EA 749 EUR
ORIGCF502254
Anti-MSI1 Mouse Monoclonal Antibody [clone: 2E9]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: MSI1
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR