Anti-SLC7A8 Mouse Monoclonal Antibody [clone: 2D4]

Supplier: OriGene
ORIGCF500631EA 749 EUR
ORIGCF500631
Anti-SLC7A8 Mouse Monoclonal Antibody [clone: 2D4]
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SLC7A8
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR