Anti-C17orf28 Mouse Monoclonal Antibody [clone: 2F4]

Supplier: OriGene
ORIGCF501311EA 749 EUR
ORIGCF501311
Anti-C17orf28 Mouse Monoclonal Antibody [clone: 2F4]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C17orf28
Clonality: Monoclonal
Clone: 2F4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR