Anti-HIBCH Mouse Monoclonal Antibody [clone: 3E7]

Supplier: OriGene
ORIGCF501320EA 770 EUR
ORIGCF501320
Anti-HIBCH Mouse Monoclonal Antibody [clone: 3E7]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HIBCH
Clonality: Monoclonal
Clone: 3E7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR