Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 5H10]

Supplier: OriGene
ORIGCF500304EA 749 EUR
ORIGCF500304
Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 5H10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: NEUROG1
Clonality: Monoclonal
Clone: 5H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR