Anti-AK1 Mouse Monoclonal Antibody [clone: 19G4]

Supplier: OriGene
ORIGCF500320EA 770 EUR
ORIGCF500320
Anti-AK1 Mouse Monoclonal Antibody [clone: 19G4]
Antibodies
Recommended Dilutions: Western Blot: 1:500-1000; Flow Cytometry: 1:100

Type: Primary
Antigen: AK1
Clonality: Monoclonal
Clone: 19G4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR