Anti-GPHN Mouse Monoclonal Antibody [clone: 3A6]

Supplier: OriGene
ORIGCF502312EA 749 EUR
ORIGCF502312
Anti-GPHN Mouse Monoclonal Antibody [clone: 3A6]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: GPHN
Clonality: Monoclonal
Clone: 3A6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR