Anti-ATP6V1F Mouse Monoclonal Antibody [clone: 6C11]

Supplier: OriGene
ORIGCF502322EA 749 EUR
ORIGCF502322
Anti-ATP6V1F Mouse Monoclonal Antibody [clone: 6C11]
Antibodies
Recommended Dilutions: Western Blot: 1:200-500; Flow Cytometry: 1:100

Type: Primary
Antigen: ATP6V1F
Clonality: Monoclonal
Clone: 6C11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR