Anti-FH Mouse Monoclonal Antibody [clone: 2A2]

Supplier: OriGene
ORIGCF500681EA 749 EUR
ORIGCF500681
Anti-FH Mouse Monoclonal Antibody [clone: 2A2]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:50-100

Type: Primary
Antigen: FH
Clonality: Monoclonal
Clone: 2A2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR