Anti-HIBCH Mouse Monoclonal Antibody [clone: 4G1]

Supplier: OriGene
ORIGCF501330EA 770 EUR
ORIGCF501330
Anti-HIBCH Mouse Monoclonal Antibody [clone: 4G1]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HIBCH
Clonality: Monoclonal
Clone: 4G1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR