Anti-HIBCH Mouse Monoclonal Antibody [clone: 3F6]

Supplier: OriGene
ORIGCF501332EA 770 EUR
ORIGCF501332
Anti-HIBCH Mouse Monoclonal Antibody [clone: 3F6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HIBCH
Clonality: Monoclonal
Clone: 3F6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR