Anti-TTC32 Mouse Monoclonal Antibody [clone: 5C12]

Supplier: OriGene
ORIGCF501341EA 770 EUR
ORIGCF501341
Anti-TTC32 Mouse Monoclonal Antibody [clone: 5C12]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: TTC32
Clonality: Monoclonal
Clone: 5C12
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR