Anti-CD19 Mouse Monoclonal Antibody [clone: 5F3]

Supplier: OriGene
ORIGCF500363EA 749 EUR
ORIGCF500363
Anti-CD19 Mouse Monoclonal Antibody [clone: 5F3]
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 5F3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR