Anti-MSI1 Mouse Monoclonal Antibody [clone: 3G4]

Supplier: OriGene
ORIGCF502370EA 749 EUR
ORIGCF502370
Anti-MSI1 Mouse Monoclonal Antibody [clone: 3G4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: MSI1
Clonality: Monoclonal
Clone: 3G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR